SHC014V
MISSION® pLKO.1-puro-UbC-TurboGFP™ Positive Control Transduction Particles
Contains a gene encoding TurboGFP, under the control of the UbC promoter
Synonym(s):
MISSION®, MISSION® Control Transduction Particles
Select a Size
Select a Size
About This Item
Quality Level
product line
MISSION®
concentration
≥1x106 VP/ml (via p24 assay)
technique(s)
capture ELISA: 106 TU/mL using p24
shipped in
dry ice
storage temp.
−70°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
To see more application data, protocols, vector maps visit sigma.com/shrna.
Application
This construct aids in interpretation of experimental design and results by providing a green fluorescent protein marker for monitoring success in transduction of your cells of interest.
Legal Information
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service