Skip to Content
Merck
All Photos(1)

Key Documents

SCP0138

Sigma-Aldrich

BQ-3020

Synonym(s):

Acetyl-(Ala 11,15)-ET 1 (6-21), Acetyl-[Ala 11,15]-endothelin 1 (6-21), BQ 3020, endothelin receptor agonist

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
$376.00
1 MG
$541.00

$376.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 μG
$376.00
1 MG
$541.00

About This Item

Empirical Formula (Hill Notation):
C96H140N20O25S1
CAS Number:
Molecular Weight:
2006.32
UNSPSC Code:
12352200
NACRES:
NA.32

$376.00


Check Cart for Availability

Request a Bulk Order

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥75%

storage condition

protect from light

UniProt accession no.

storage temp.

−20°C

SMILES string

S(CC[C@H](NC(=O)[C@@H](NC(=O)C)CC(C)C)C(=O)N[C@@H](CC(=O)O)C(=O)N[C@@H](CCCCN)C(=O)NCC(=O)N[C@@H](C)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](Cc5ccc(cc5)O)C(=O)N[C@@H](Cc4ccccc4)C(=O)N[C@@H](C)C(=O)N[C@@H](Cc3[nH]cnc3)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CC(=O)O)C(=O)N

InChI

1S/C93H136N20O23S/c1-15-51(9)77(91(133)110-72(93(135)136)40-58-44-96-62-27-21-20-26-61(58)62)113-92(134)78(52(10)16-2)112-89(131)71(43-75(119)120)108-85(127)66(37-49(5)6)105-87(129)69(41-59-45-95-47-98-59)104-79(121)54(12)100-83(125)67(38-56-24-18-17-19-25-56)106-86(128)68(39-57-29-31-60(115)32-30-57)109-90(132)76(50(7)8)111-80(122)53(11)99-73(116)46-97-81(123)63(28-22-23-34-94)102-88(130)70(42-74(117)118)107-82(124)64(33-35-137-14)103-84(126)65(36-48(3)4)101-55(13)114/h17-21,24-27,29-32,44-45,47-54,63-72,76-78,96,115H,15-16,22-23,28,33-43,46,94H2,1-14H3,(H,95,98)(H,97,123)(H,99,116)(H,100,125)(H,101,114)(H,102,130)(H,103,126)(H,104,121)(H,105,129)(H,106,128)(H,107,124)(H,108,127)(H,109,132)(H,110,133)(H,111,122)(H,112,131)(H,113,134)(H,117,118)(H,119,120)(H,135,136)/t51-,52-,53-,54-,63-,64-,65-,66-,67-,68-,69-,70-,71-,72-,76-,77-,78-/m0/s1

InChI key

NUAGCEFLVCODRQ-KETPNHLCSA-N

Gene Information

human ... EDN1(1906)

Amino Acid Sequence

Ac-Leu-Met-Asp-Lys-Glu-Ala-Val-Tyr-Phe-Ala-His-Leu-Asp-Ile-Ile-Trp

Biochem/physiol Actions

BQ-3020, a linear endothelin analog, is a selective ETB endothelin receptor agonist.[1][2]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel radioligand [125I]BQ-3020 selective for endothelin (ETB) receptors.
Ihara M
Life Sciences, 51(6) (1992)
Q T Nguyen et al.
Circulation, 104(17), 2075-2081 (2001-10-24)
Some controversy exists as to the effects of endothelin (ET) receptor antagonism on long-term post-myocardial infarction (MI) evolution, particularly as it relates to the timing of the intervention after MI (<24 hours versus 10 days). Sham rats and rats surviving
Phuong N Quang et al.
Pain, 149(2), 254-262 (2010-03-09)
Endothelin-1 (ET-1) produced by various cancers is known to be responsible for inducing pain. While ET-1 binding to ETAR on peripheral nerves clearly mediates nociception, effects from binding to ETBR are less clear. The present study assessed the effects of
Peter Johnström et al.
Experimental biology and medicine (Maywood, N.J.), 231(6), 736-740 (2006-06-03)
The endothelin (ET) receptor system has been shown to play a role in a number of vascular diseases. We have synthesized 18F-and 11C-labeled radioligands to enable in vivo imaging of the fundamental processes involved in ET receptor pharmacology in normal
A. Bjorklund
Peptides (New York, NY, United States) (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service