Skip to Content
Merck
All Photos(1)

Key Documents

SCP0134

Sigma-Aldrich

Endothelin Antagonist PD 142893

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$269.00
5 MG
$929.00

$269.00


Check Cart for Availability


Select a Size

Change View
1 MG
$269.00
5 MG
$929.00

About This Item

Empirical Formula (Hill Notation):
C50H65N7O10
Molecular Weight:
924.09
UNSPSC Code:
12352200
NACRES:
NA.32

$269.00


Check Cart for Availability

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥85%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Acl-Dif-Leu-Asp-Ile-Ile-Trp

Application

PD 142-893 is used as a nonselective endothelin ET(A)/ET(B) receptor antagonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chumpon Wilasrusmee et al.
Surgery, 134(2), 384-389 (2003-08-30)
We have previously shown that endothelial injury by cyclosporin A (CyA) is associated with an increased endothelin-1 (ET-1) release. We now sought to determine, in an animal model of angiogenesis, if inhibiting the effect of ET-1 on endothelial cells (ECs)
Ashley M Stokes et al.
Canadian journal of veterinary research = Revue canadienne de recherche veterinaire, 70(3), 197-205 (2006-07-21)
The goals of this study were to determine the concentration-response (C-R) relationship of endothelin-1 (ET-1), compare 2 ET-receptor antagonists and determine the antagonist concentrations that block the vasomotor effects of ET-1, and compare the effectiveness of ET-1 and previously studied
Sebastien Faure et al.
Journal of vascular research, 43(1), 19-26 (2005-10-29)
The effect of angiotensin IV (AngIV) was studied in freshly isolated rat basilar arteries (BAs) perfused at a constant rate. AngIV had no effect on basal BA perfusion pressure, but induced a marked concentration-dependent contraction in vessels precontracted by a
Alexandra Madsen et al.
Journal of molecular and cellular cardiology, 154, 115-123 (2021-02-15)
The role of DNA methylation in cardiomyocyte physiology and cardiac disease remains a matter of controversy. We have recently provided evidence for an important role of DNMT3A in human cardiomyocyte cell homeostasis and metabolism, using engineered heart tissue (EHT) generated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service