Skip to Content
Merck
All Photos(1)

Documents

SCP0058

Sigma-Aldrich

Bradykinin Peptide

≥95% (HPLC), lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C59H89N17O14
Molecular Weight:
1260.44
UNSPSC Code:
12352202
NACRES:
NA.32

product name

Bradykinin Ile-Ser, T-Kinin,

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥65%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Ile-Ser-Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg

Application

Bradykinin, a nonapeptide, is a potent endothelium-dependent vasodilator that operates via bradykinin receptors B1 and B2. Bradykinin Ile-Ser (T-Kinin) has a biological profile similar but not identical to bradykinin which is more resistant to degradation by kininase angiotensin converting enzyme (ACE). Bradykinin Ile-Ser is used to differentiate bradykinin receptor specificity and function.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Bernhold Brechter et al.
Regulatory peptides, 103(1), 39-51 (2001-12-12)
Bradykinin receptor subtypes linked to prostaglandin release have been assessed in a human osteosarcoma cell line with osteoblastic phenotype (MG-63). Bradykinin (BK; 1 micromol/l) caused a burst of prostaglandin E(2) release that was maximal at 10 min. When the effect
D G Sawutz et al.
European journal of pharmacology, 227(3), 309-315 (1992-11-02)
[3H]Bradykinin binds to intact human IMR-90 fetal lung fibroblasts in a time and dose-dependent manner. Binding equilibrium was attained by 120 minutes at 4 degrees C. [3H]Bradykinin binding was saturable; Scatchard analysis of saturation binding data demonstrated a single binding
Prashant Desai et al.
European journal of medicinal chemistry, 37(2), 135-146 (2002-02-23)
T-kinin (Ile-Ser-BK) is related to BK in its biological profile, but unlike BK, is more resistant to the action of ACE. A detailed NMR and molecular modeling study of T-kinin has been carried out in three diverse media: water (pH
Clare Davis et al.
European journal of pharmacology, 450(2), 123-130 (2002-09-11)
T-kinin and its putative carboxypeptidase product des-Arg(11)-T-kinin are members of the kinin family that are unique to the rat. Primary cultures of rat bladder smooth muscle cells were used to investigate the pharmacology of these peptides. Calcium imaging experiments showed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service