Skip to Content
Merck
All Photos(1)

Key Documents

SCP0025

Sigma-Aldrich

Gamma Secretase Inhibitor Peptide

≥95% (HPLC), lyophilized powder

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$156.00

$156.00


Estimated to ship on31 May 2025



Select a Size

Change View
5 MG
$156.00

About This Item

Empirical Formula (Hill Notation):
C20H20N2O4
Molecular Weight:
352.38
UNSPSC Code:
12352202
NACRES:
NA.32

$156.00


Estimated to ship on31 May 2025


Product Name

APP γ Secretase Inhibitor,

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥75%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

3-5 difluorophenacetyl-Ala-S-Phenylglycine-Methylester

Application

Gamma-secretase is a protein complex containing presenilin, nicastrin, APH-1, and PEN-2. 3-5 difluorophenacetyl-Ala-S-Phenylglycine-Methylester is used as an amyloid precursor protein (APP) gamma secretase inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service