Skip to Content
Merck
All Photos(1)

Key Documents

SCP0020

Sigma-Aldrich

[Sar1]-Angiotensin II

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$269.00

$269.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$269.00

About This Item

Empirical Formula (Hill Notation):
C49H71N13O10
Molecular Weight:
1002.17
UNSPSC Code:
12352209
NACRES:
NA.32

$269.00


Check Cart for Availability

Request a Bulk Order

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥65%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Sar-Arg-Val-Tyr-Ile-His-Pro-Phe

Application

Angiotensins are peptide hormone components of the renin-angiotensin system involved in the regulation of blood vessel constriction and aldosterone release. Angiotensin I, a decapeptide derived from angiotensinogen, is a converted to Angiotensin II by the removal of the two C-terminal amino acids. Various analogues of Angiotensins I and II made by amino acid substitution are used in structure and sequence studies to analyze and differentiate the functions of natural angiotensins.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kar Neng Lai et al.
Contributions to nephrology, 170, 135-144 (2011-06-11)
Diabetic nephropathy (DN) is a leading cause of end-stage renal disease in developed countries where type 2 diabetes mellitus has reached epidemic proportions. Although the exact pathogenesis of DN is not fully understood and is likely diverse in nature, there
Vani X Oliveira et al.
Regulatory peptides, 172(1-3), 1-7 (2011-07-27)
Angiotensin II (AII) is the active octapeptide product of the renin enzymatic cascade, which is responsible for sustaining blood pressure. In an attempt to establish the AII-receptor-bound conformation of this octapeptide, we designed conformationally constrained analogues by scanning the entire
Novel concept in the mechanism of injury and protection of gastric mucosa: role of Renin-Angiotensin system and active metabolites of Angiotensin.
Brzozowski T, Ptak-Belowska A, et al.
Current Medicinal Chemistry, 16, 55-62 (2012)
Mauricio Sendeski et al.
Radiology, 251(3), 697-704 (2009-04-16)
To determine whether a type of contrast medium (CM), iodixanol, modifies outer medullary descending vasa recta (DVR) vasoreactivity and nitric oxide (NO) production in isolated microperfused DVR. Animal handling conformed to the Animal Care Committee Guidelines of all participating institutions.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service