Skip to Content
Merck
All Photos(1)

Key Documents

SCP0002

Sigma-Aldrich

Acetyl-Amyloid β 25-35

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$134.00
5 MG
$460.00

$134.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$134.00
5 MG
$460.00

About This Item

Empirical Formula (Hill Notation):
C47H83N13O15S1
Molecular Weight:
1102.31
UNSPSC Code:
12352202
NACRES:
NA.32

$134.00


Check Cart for Availability

Request a Bulk Order

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥70%

storage condition

protect from light

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... APP(351)

Amino Acid Sequence

Ac-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met

General description

Amyloid-β (Aβ) peptides are a major component of the senile plaques characteristic of the Alzheimer brain.[1] It is a type- I transmembrane protein. The amyloid β precursor protein is cleaved to amyloid peptides 36-43 amino acids in length. Aβ (25-35) represents the region that aggregates during plaque formation and is a functional domain implicated in both neurotrophic and neurotoxic effects. The gene encoding amyloid-β is localized on human chromosome 21. Acetylation may improve stability in in vitro and in vivo assays.[2][3]

Application

Acetyl-Amyloid β 25-35 has been used to prepare rat models with Alzheimer′s disease.[4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Conformational Dynamics of Specific A? Oligomers Govern Their Ability To Replicate and Induce Neuronal Apoptosis.
Dean DN
Biochemistry, 55(15), 2238-2250 (2016)
Monomeric Amyloid Beta Peptide in Hexafluoroisopropanol Detected by Small Angle Neutron Scattering.
Zhang-Haagen B
PLoS ONE, 11(2), e0150267-e0150267 (2016)
Effect of yizhitongxuan decoction on learning and memory ability, Gaq/11 expression and Na(+)-K(+)-ATP enzyme activity in rat models of Alzheimer's disease.
Teng J
Journal of Traditional Chinese Medicine = Chung i Tsa Chih Ying Wen Pan / Sponsored by All-China Association of Traditional Chinese Medicine, Academy of Traditional Chinese Medicine, 34(4), 470-476 (2014)
Yili Wu et al.
Scientific reports, 6, 22460-22460 (2016-03-05)
Down syndrome (DS), caused by trisomy of chromosome 21, is one of the most common genetic disorders. Patients with DS display growth retardation and inevitably develop characteristic Alzheimer's disease (AD) neuropathology, including neurofibrillary tangles and neuritic plaques. The expression of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service