Skip to Content
Merck
All Photos(1)

Key Documents

SCJ4600039

Sigma-Aldrich

CF 633 succinimidyl ester

suitable for fluorescence, ≥90% (TLC)

Synonym(s):

Fluorescent dye, Succinimidyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

1 μMOL
$534.00

$534.00


Available to ship on23 April 2025Details



Select a Size

Change View
1 μMOL
$534.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$534.00


Available to ship on23 April 2025Details


Quality Level

Assay

≥90% (TLC)

form

solid

mol wt

~821 g/mol (free acid)

solubility

DMF: soluble
DMSO: soluble
H2O: soluble

ε (extinction coefficient)

100000 at 630 nm

absorbance ratio

A280/630 nm 0.48

fluorescence

λex 633 or 635 nm (Flow cytometry laser line)
λex 633 or 635 nm (Microscopy laser line)
λex 630 nm; λem 650 nm

suitability

suitable for fluorescence

shipped in

wet ice

storage temp.

−20°C

General description

Far-red fluorescent dyes offer the advantage of ultra sensitive detection because background signal due to autofluorescence in most biological samples is minimal in this spectral region. For many years, the cyanine dye Cy®5 has been the dye of choice for such detection. More recently, Alexa Fluor 647 has been developed as a better alternative by having brighter fluorescence and higher photostability. Despite the improvement, Alexa Fluor 647 still lacks sufficient photostability required for many demanding applications. On the other hand, while Alexa Fluor 633 is photostable, its fluorescence on proteins is very weak. In fact, it has been a challenging task for dye chemists to develop a far-red fluorescent dye that is both highly fluorescent and photostable for protein and nucleic acid labeling. Using new chemistry, scientists at Biotium have successfully developed a series of far-red and near-IR dyes to overcome these challenges. With its absorption peak at 630 nm, CF633 can be optimally excited by the 633 nm He-Ne laser or the 635 nm red diode laser with emission maximum at 650 nm. Despite its shorter emission wavelength, which often is not favored by a Cy®5-based detection window on most instruments, CF633 is still significantly brighter than Alexa Fluor 647. However, for detections using non-laser-based excitation and a Cy®5 filter set, the full advantage of CF633 will not be realized. In this case, we recommend CF640R, another outstanding far-red CF dye that is spectrally closer to Cy®5 or other similar cyanine-based dyes. The most important advantage of CF633 and other rhodamine-based far-red CF dyes are their unmatched photostability. The combination of superior brightness and photostability makes far-red CF dyes ideal choices for detections in the long wavelength region on a variety of instruments.

Features and Benefits

1. Yields the brightest anibody conjugates among spectrally similar dyes when excited by the 633 nm He-Ne laser or the 635 nm red diode laser. 2. Far more photostable than Alexa Fluor647. 3. Highly water-soluble.

Preparation Note

Protect from light and moisture

Reconstitution

Dissolve in DMSO

Legal Information

This product is distributed by Sigma-Aldrich Co. under the authorization of Biotium, Inc. This product is covered by one or more US patents and corresponding patent claims outside the US patents or pending applications owned or licensed by Biotium, Inc. including without limitation: 12/334,387; 12/607,915; 12/699,778; 12/850,578; 61/454,484. In consideration of the purchase price paid by the buyer, the buyer is hereby granted a limited, non-exclusive, non-transferable license to use only the purchased amount of the product solely for the buyer′s own internal research in a manner consistent with the accompanying product literature. Except as expressly granted herein, the sale of this product does not grant to or convey upon the buyer any license, expressly, by implication or estoppel, under any patent right or other intellectual property right of Biotium, Inc. Buyer shall not resell or transfer this product to any third party, or use the product for any commercial purposes, including without limitation, any diagnostic, therapeutic or prophylactic uses. This product is for research use only. Any other uses, including diagnostic uses, require a separate license from Biotium, Inc. For information on purchasing a license to use this product for purposes other than research, contact Biotium, Inc., 3159 Corporate Place, Hayward, CA 94545, Tel: (510) 265-1027. Fax: (510) 265-1352. Email: [email protected].
ALEXA FLUOR is a trademark of Life Technologies
CF is a trademark of Biotium, Inc.
Cy is a registered trademark of Cytiva

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Targeted detection of murine colonic dysplasia in vivo with flexible multispectral scanning fiber endoscopy.
Miller, S.J., et al.
Journal of Biomedical Optics, 2, 021103-021103 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service