Skip to Content
Merck
All Photos(2)

Key Documents

SBR00055

Sigma-Aldrich

Polar Metabolites QC Mix

suitable for LC-MS

Sign Into View Organizational & Contract Pricing

Select a Size

0.2 ML
$416.00

$416.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
0.2 ML
$416.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.28

$416.00


Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

packaging

pkg of 1 vial

suitability

suitable for LC-MS

storage temp.

2-8°C

General description

In order to assess any variations in LC-MS-based metabolomics analysis data, it is recommended to add an additional QC (quality control) sample at the beginning of every analytical experiment batch. It is recommended that QC be injected every 4-10 injections into the workflow steps.[1][2] The QC sample evaluates any drifting phenomenon like intensity values, ion suppression, or any changes in the retention time of the material’s peaks in the LC-MS data. The Polar Metabolites QC Mix is a ready to use solution for mass spectroscopy workflows. By utilizing the QC mix, the user will be able to assess drifting and ion suppression phenomenon. The Polar Metabolites QC Mix is comprised of 8 components (Uridine, Adenosine, Nicotinic acid, Biotin, Phenylacetylglutamine, L-phenylalanine, Cytidine and L-Tryptophan) which covers ~ 100-500 Da mass range.

Features and Benefits

  • The mix was designed for ESI (+/-) detection.
  • The mixture comes as a ready to use solution(~9:1 ACN:10mM ammonium formate, inwater) format in crimp top (Silicone/PTFE liner) amber vial, which is suitable for use with most of the LC-MS autosamplers.
  • The recommended storage temperature of the mixture is 2-8 °C, thawing of the sample is not required.

Suitability

The Polar Metabolites QC Mix is suitable for mass spectroscopy

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service