Skip to Content
Merck
All Photos(1)

Key Documents

SBR00051

Sigma-Aldrich

Trizma® hydrochloride solution

pH 8.5, DNase and Microbial DNA free, 10 mM

Synonym(s):

Trizma® hydrochloride, TRIS HCl, TRIS hydrochloride, Tris(hydroxymethyl)aminomethane hydrochloride, Tromethane hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$131.00

$131.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$131.00

About This Item

Linear Formula:
NH2C(CH2OH)3 · HCl
CAS Number:
Molecular Weight:
157.60
Beilstein:
3675235
MDL number:
UNSPSC Code:
12161700
NACRES:
NA.25

$131.00


Check Cart for Availability

Request a Bulk Order

Quality Level

concentration

10 mM

pH

8.5

useful pH range

7.0-9.0

pKa (25 °C)

8.1

storage temp.

2-8°C

SMILES string

Cl.NC(CO)(CO)CO

InChI

1S/C4H11NO3.ClH/c5-4(1-6,2-7)3-8;/h6-8H,1-3,5H2;1H

InChI key

QKNYBSVHEMOAJP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

10 mM Trizma® hydrochloride (pH 8.5) is a widely used buffer in molecular biology applications such as nucleic acid extraction and purification. No mixing or pH adjustment is necessary. Guaranteed accuracy for this buffer is ± 0.2 pH units at 25°C.

Application

Trizma® hydrochloride solution can be used:
  • As an elution buffer after PCR cleanup
  • As an elution buffer after DNA extraction

Preparation Note

Allow the buffer to reach room temperature before use.

Other Notes

Prepared with BioUltra Trizma Base for molecular biology in 18 megaohm UV irradiated water. pH-adjusted and 0.2 μm filtered. The buffer was tested by 35 cycles of PCR for prokaryotic and eukaryotic DNA contamination. To keep the buffer contamination-free, it is recommended to use gloves and barrier-tip pipettes for dispensing. Between uses, keep the bottle closed and refrigerated, away from areas where DNA samples and PCR products may be stored.
The pH values of all buffers are temperature and concentration dependent. For Tris buffers, pH increases about 0.03 unit per °C decrease in temperature, and decreases 0.03-0.05 unit per ten-fold dilution. For precise applications, use a carefully calibrated pH meter with a glass/calomel combination electrode.

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service