Skip to Content
Merck
All Photos(1)

Key Documents

SBR00017

Sigma-Aldrich

Puromycin aminonucleoside Ready Made Solution

10 mg/mL in water

Synonym(s):

3′-Amino-3′-deoxy-N6,N6-dimethyladenosine

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$255.00

$255.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$255.00

About This Item

Empirical Formula (Hill Notation):
C12H18N6O3
CAS Number:
Molecular Weight:
294.31
UNSPSC Code:
51281912
NACRES:
NA.85

$255.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98%

Quality Level

form

liquid

concentration

10 mg/mL in water

antibiotic activity spectrum

Gram-positive bacteria
neoplastics
parasites

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

N[C@H]1[C@@H](O)[C@H](N(C=N2)C3=C2C(N(C)C)=NC=N3)O[C@@H]1CO

InChI

1S/C12H18N6O3/c1-17(2)10-8-11(15-4-14-10)18(5-16-8)12-9(20)7(13)6(3-19)21-12/h4-7,9,12,19-20H,3,13H2,1-2H3

InChI key

RYSMHWILUNYBFW-UHFFFAOYSA-N

Biochem/physiol Actions

Puromycin aminonucleoside is an aminonucleoside derivative of the known antibiotic puromycin. Puromycin aminonucleoside has been used in nephrology research, studying focal and segmental glomerulosclerosis, and in the induction of nephrosis in rats.[1][2][3] Rats with puromycin aminonucleoside-induced nephrotic syndrome were used to study the excretion of sodium and NOx metabolites.[4]

Puromycin aminonucleoside has also been used to probe endothelial glycosaminoglycan synthesis in cultured glomerular endothelial cells and their relation to cell permeability.[5] A puromycin aminonucleoside nephrosis model of rat glomerular disease was also used to study the role of the neuron-specific ubiquitin C-terminal hydrolase protein gene product 9.5 (PGP 9.5).[6]

Preparation Note

Puromycin aminonucleoside solution is provided at 10 mg/mL in water and could be further diluted in aqueous buffers to a working concentration of 10 μg/mL (1:1000).

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Agnes B Fogo
Seminars in nephrology, 23(2), 161-171 (2003-04-22)
Glomerulosclerosis in a heterogeneous pattern, ie, focal and segmental glomerulosclerosis (FSGS), is a common endpoint in a variety of settings, including idiopathic FSGS, and scarring secondary to other renal or systemic diseases. These different causes contribute to the diverse clinical
I Shirato et al.
Journal of the American Society of Nephrology : JASN, 11(12), 2381-2386 (2000-11-30)
Parietal epithelial cells (PEC) of Bowman's capsules cover the inner aspect of Bowman's capsules and are believed to contribute to extracapillary lesions of glomerulonephritis such as crescent formation. In glomerular research including cell culture experiments and pathology, differentiation between PEC
Alicia A McDonough et al.
Current opinion in nephrology and hypertension, 12(5), 533-541 (2003-08-16)
The proximal tubule sodium/hydrogen exchanger continuously reabsorbs the bulk of the filtered sodium, controlling salt delivery to the distal nephron which is critical for tubuloglomerular feedback autoregulation and for fine control of salt excretion in the distal nephron. This review
Jenny Sörensson et al.
American journal of physiology. Renal physiology, 284(2), F373-F380 (2002-10-22)
It has been suggested that proteinuria is caused by alterations of the charge selectivity of the basement membrane and/or the epithelial cell layer (podocytes). However, recent findings suggest that the endothelial luminal surface coat, consisting of proteoglycans with their connected
Marta Bermejo-Jambrina et al.
Frontiers in immunology, 11, 2010-2010 (2020-09-15)
Dendritic cells (DCs) possess intrinsic cellular defense mechanisms to specifically inhibit HIV-1 replication. In turn, HIV-1 has evolved strategies to evade innate immune sensing by DCs resulting in suboptimal maturation and poor antiviral immune responses. We previously showed that complement-opsonized

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service