SBPVR3
SB PREDIVEZ™ Reagent Kit for MRP2, MRP3, and MRP5
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
recombinant
expressed in human cells
form
liquid
shipped in
dry ice
storage temp.
−70°C
General description
SOLVO Biotechnology offers ready-to-use assay kits for efflux and uptake transporters. SOLVO′s PREDIVEZ Vesicular Transport Assay Kits are suitable for efflux transporter inhibition studies. The kits are designed to test the inhibitory effect of one or more test compounds on the vesicular transport of a probe substrate. In most cases a fluorescent probe substrate is included. Kits contain all reagents and solutions necessary to conduct the drug-transporter interaction assay of need.
Application
The PREDIVEZ Vesicular Transport Kit is a simple and powerful tool to investigate drug transporter interactions. It provides information on any interaction between the ABC transporter and the drug candidate that would affect the transport of the fluorescent or radiolabeled or cold.
PREDIVEZ vesicular transport kits contains buffers, probe substrate, and reference inhibitors needed to run vesicular transport assays (sufficient for 300 wells/kit –equivalent to existing 9-cpd PV kits). Assay protocol, calculation template, and example data file available on SOLVO website.
Transport membranes and control membranes are sold separately. This PREDIVEZ vesicular transport kit requires the transporter membranes and corresponding control membrane below:
SB-MRP2-Sf9-VT MRP2 human membrane vesicles part number SBVT05-1EA and corresponding control membrane SB-beta-gal-Sf9-CTRL part number SBCT03-1EA
SB-MRP2-HEK293-VT MRP2 human membrane vesicles part number SBVT14-1EA and corresponding control membrane SB-HEK293-Mock-CTRL part number SBCT06-1EA
SB-MRP3-Sf9-VT MRP3 human membrane vesicles part number SBVT06-1EA and corresponding control membrane SB-beta-gal-Sf9-CTRL part number SBCT03-1EA
SB-MRP3-HEK293-VT MRP3 human membrane vesicles part number SBVT15-1EA and corresponding control membrane SB-HEK293-Mock-CTRL part number SBCT06-1EA
SB-MRP5-HEK293-VT MRP5 human membrane vesicles part number SBVT08-1EA and corresponding control membrane SB-HEK293-Mock-CTRL part number SBCT06-1EA
PREDIVEZ vesicular transport kits contains buffers, probe substrate, and reference inhibitors needed to run vesicular transport assays (sufficient for 300 wells/kit –equivalent to existing 9-cpd PV kits). Assay protocol, calculation template, and example data file available on SOLVO website.
Transport membranes and control membranes are sold separately. This PREDIVEZ vesicular transport kit requires the transporter membranes and corresponding control membrane below:
SB-MRP2-Sf9-VT MRP2 human membrane vesicles part number SBVT05-1EA and corresponding control membrane SB-beta-gal-Sf9-CTRL part number SBCT03-1EA
SB-MRP2-HEK293-VT MRP2 human membrane vesicles part number SBVT14-1EA and corresponding control membrane SB-HEK293-Mock-CTRL part number SBCT06-1EA
SB-MRP3-Sf9-VT MRP3 human membrane vesicles part number SBVT06-1EA and corresponding control membrane SB-beta-gal-Sf9-CTRL part number SBCT03-1EA
SB-MRP3-HEK293-VT MRP3 human membrane vesicles part number SBVT15-1EA and corresponding control membrane SB-HEK293-Mock-CTRL part number SBCT06-1EA
SB-MRP5-HEK293-VT MRP5 human membrane vesicles part number SBVT08-1EA and corresponding control membrane SB-HEK293-Mock-CTRL part number SBCT06-1EA
Legal Information
PREDIVEZ is a trademark of Solvo Biotechnology
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A
Storage Class Code
8A - Combustible corrosive hazardous materials
Flash Point(F)
188.6 °F
Flash Point(C)
87 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service