Skip to Content
Merck
All Photos(1)

Key Documents

SAMMS2HYG

Sigma-Aldrich

MS2-P65-HSF1-Hygromycin SAM CRISPRa Helper Construct 2 Plasmid DNA

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$619.00

$619.00


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
10 μG
$619.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.51

$619.00


Check Cart for Availability

Request a Bulk OrderRequest more information

packaging

vial of 20 μL

concentration

500 ng/μL in TE buffer; DNA (10μg of plasmid DNA)

application(s)

CRISPR

shipped in

dry ice

storage temp.

−20°C

General description

This product is a lentiviral plasmid that utilizes the EF1 alpha promoter to drive expression of MS2-P65-HSF1 and hygromycin resistance cassette linked by a 2A peptide (EF1a-MS2-P65-HSF1-2A-Hygromycin) allowing for easy selection following successful transfection or transduction. Use Sigma′s lentiviral MS2-P65-HSF1 plasmid for generation of lentiviral particles and efficient production of stable cell lines expressing MS2-P65-HSF1 for CRISPR based transcipritonal activation and genome wide gain of function screening. Sigma′s lenti-MS2-P65-HSF1 plasmid is one part of a three part CRISPR system with individual dCas9-VP64, MS2-p65-HSF1 and gRNA expression vectors.

To order gRNA in any format click here

Application

Functional Genomics/Target Validation
  • Unbiased forward genetic screening
  • Strong transcriptional activation in multiple cell lines
  • Manufacture of MS2-p65-HSF1 expressing Lentiviral Particles

Features and Benefits

  • Highly specific and highly active
  • Sequence verified high purity plasmid DNA
  • Activates genes through transcriptional activation rather than cDNA based overexpression

Learn more about SAM CRISPR Activators at SigmaAldrich.com/CRISPRa

Principle

CRISPR/Cas systems are employed by bacteria and archaea as a defense against invading viruses and plasmids. Recently, the type II CRISPR/Cas system from the bacterium Streptococcus pyogenes has been engineered to function in eukaryotic systems using two molecular components: a single Cas9 protein and a non-coding guide RNA (gRNA). The Cas9 endonuclease can be rendered inactive (dCas9) with mutations to the two protein domains, RuvC and HnH (D10A and H840A respectively), which are responsible for nuclease activity. The nuclease deficient protein can then be fused with the transcriptional activator VP64 and used in conjunction with a guide RNA modified with MS2 RNA aptamers that function to recruit the additional transcriptional coactivators p65 and HSF1. The assembled SAM complex is then used as a cargo delivery system to target gene promoters, enabling site-specific transcriptional activation of the gene of interest[1]

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genome-scale transcriptional activation by an engineered CRISPR-Cas9 complex
Konermann S, et al
Nature, 517, 83-588 (2015)
Genome-scale CRISPR-Cas9 Knockout and Transcriptional Activation Screening.
Joung, S. et al.
Nature Protocols, 12, 828-863 (2017)
Julia Joung et al.
Nature protocols, 12(4), 828-863 (2017-03-24)
Forward genetic screens are powerful tools for the unbiased discovery and functional characterization of specific genetic elements associated with a phenotype of interest. Recently, the RNA-guided endonuclease Cas9 from the microbial CRISPR (clustered regularly interspaced short palindromic repeats) immune system

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service