SAE0219
Mucin Degradation Enzyme
for preparative digestions, powder
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Quality Level
form
powder
shelf life
2 yr (retest)
storage condition
protect from light
shipped in
dry ice
storage temp.
-10 to -25°C
General description
Mucin Degradation Enzyme is a metalloprotease useful for digesting heavily O-glycosylated mucin proteins. This product is made for preparative scale digestions of viscous material that is dense with mucin-type glycoproteins.
Application
For use as a sample preparation tool in research and discovery. Used for proteolytic degradation of mucin-dense tissues to allow for downstream processing.
Biochem/physiol Actions
The Mucin Degradation enzyme is a mutated form of a protease that specifically recognizes densely glycosylated mucins and mucin-domains, and proteolytically cleaves the peptide backbone. In nature this presumably enhances the virulence of the pathogenic microbe, but in the laboratory is a useful way of accessing molecular structures in tissues normally shielded by mucins.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service