Skip to Content
Merck
All Photos(1)

Key Documents

SAE0202

Sigma-Aldrich

Mucinase StcE

mass spec suitable, from EHEC, recombinant, powder

Synonym(s):

Mucin-specific protease StcE, secreted protease of C1 esterase inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$581.00
5 X 100 μG
$1,830.00

$581.00


Check Cart for Availability


Select a Size

Change View
100 μG
$581.00
5 X 100 μG
$1,830.00

About This Item

UNSPSC Code:
12352204
NACRES:
NA.54

$581.00


Check Cart for Availability

Quality Level

shipped in

wet ice

storage temp.

-10 to -25°C

General description

Mucinase StcE is a metalloprotease that can be used to digest densely O-glycosylated mucins. The consensus sequence for cleavage is S/T*-X-S/T, where the cleavage happens on the amino side of the 2nd Serine/Threonine, and X can be any amino acid, or none though with lower efficiency. The P2 position (*) is found to be always occupied in cleavage products, and occupancy of both P1 and P1’ positions do not interfere with cleavage. While Mucinase StcE is active against Mucins and other mucin-domain containing glycoproteins such as CD43 or PSGL-1, it is not active against O-glycoproteins outside of the Mucin family, such as fetuin or GspB736flag1.

Application

For use as a tool in biochemical research and biomarker discovery, particularly focused on glycobiology, glycoproteomics, glyco-oncology. Shown to be useful in LC-MS/MS workflows

Biochem/physiol Actions

Mucins are heavily O-glycosylated proteins found in many tissues such as mucosal membranes and intestinal linings. One of its functions is as a protective layer for the epithelial tissue. The pathogenic bacteria enterohemorrhagic Escherichia coli (EHEC) is known to attack these tissues by enzymatic means. One of the enzymes found to be involved is a protease called secreted protease of C1 esterase inhibitor (StcE) (a.k.a. Mucinase StcE). This enzyme specifically recognizes densely glycosylated mucins and mucin-domains, and proteolytically cleaves the peptide backbone, presumably enhancing the virulence of the pathogenic microbe.

Features and Benefits

  • Protease highly selective to mucins, but promiscuous within mucin family.
  • Tool for O-linked glycopeptide release
  • Configured in packages useful for approximately 40 samples
  • His-Tag fusion for easy purification and isolation

Storage and Stability

The product retains activity for at least 2 years when stored lyophilized at -20°C

Other Notes

Solution of StcE can be prepared by reconstitution of the lyophilized 20 µg vial in 100 µL of water or PBS. The resuspended protein solution is stable for 2 weeks at 2-8 °C. For longer storage, aliquot the protein solution and store at -20 °C. The enzyme solution should be protected from direct light.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid and Sensitive Detection of Native GlycoRNAs
Hemberger, H.; Chai, P.; Lebedenko, C. G.; Caldwell, R. M.; George, B. M.; Flynn, R. A
bioRxiv : the preprint server for biology (2023)

Articles

Pretreatment with Mucinase StcE increases glycopeptide identification from mucin samples, enhancing sample preparation efficiency for glycopeptide analysis.

Protocols

Explore mass spectrometry analysis of glycans for glycomic & glycoproteomic neutral & acidic glycan analysis. See a general mass spec glycan analysis procedure.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service