Skip to Content
Merck
All Photos(2)

Key Documents

SAE0062

Sigma-Aldrich

Lipodisq®

Styrene:Maleic Anhydride Copolymer 2:1, Pre-hydrolyzed

Synonym(s):

Lipodisq Reagent, Lipodisq Solution

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$634.00
500 MG
$2,230.00

$634.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$634.00
500 MG
$2,230.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.25

$634.00


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

storage temp.

room temp

General description

Lipodisq nanoparticles are usually made up of styrene maleic acid (SMA) copolymer and phospholipids. It has almost similar structure to that of nanodiscs.[1] Lipodisq nanoparticles are considered as a good membrane mimetic system for the structural studies of membrane proteins.[2]

Application

Lipodisq reagents are novel polymers derived from styrene and maleic acid. Lipodisq polymers are capable of forming nanosized lipid-based discoidal particles capable of incorporating membrane proteins. The Lipodisq technology can solubilize commonly used lipids such as dimyristoylphosphatidylcholine (DMPC) without the use of detergents. Lipodisqs typically have a diameter of 9-10 nm at pH 7.4.

The Lipodisq technology can solubilize commonly used lipids such as dimyristoylphosphatidylcholine (DMPC) without the use of detergents.

Functional pH range: 5.5 - 8.0

Biochem/physiol Actions

Lipodisq nanoparticles are considered as a good membrane mimetic system for the structural studies of membrane proteins.[2]

Packaging

100, 500 mg in poly bottle

Legal Information

Lipodisq is a registered trademark of Malvern Cosmeceutics Limited

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of the structure of lipodisq nanoparticles in the presence of KCNE1 by dynamic light scattering and transmission electron microscopy
Zhang R, et al.
Chemistry and Physics of Lipids, 203, 19-23 (2017)
Characterization of KCNE1 inside lipodisq nanoparticles for EPR spectroscopic studies of membrane proteins
Sahu I D, et al.
The Journal of Physical Chemistry B, 121(21), 5312-5321 (2017)
Lina Zhu et al.
PeerJ, 10, e13381-e13381 (2022-05-10)
Smoothened (SMO) protein is a member of the G protein-coupled receptor (GPCR) family that is involved in the Hedgehog (Hh) signaling pathway. It is a putative target for treating various cancers, including medulloblastoma and basal cell carcinoma (BCC). Characterizing membrane
Juan Francisco Bada Juarez et al.
Biochimica et biophysica acta. Biomembranes, 1862(3), 183152-183152 (2019-12-18)
Dopamine receptors (DRs) are class A G-Protein Coupled Receptors (GPCRs) prevalent in the central nervous system (CNS). These receptors mediate physiological functions ranging from voluntary movement and reward recognition to hormonal regulation and hypertension. Drugs targeting dopaminergic neurotransmission have been
Juan Francisco Bada Juarez et al.
Chemistry and physics of lipids, 221, 167-175 (2019-04-04)
Nanoparticles assembled with poly(styrene-maleic acid) copolymers, identified in the literature as Lipodisq, SMALPs or Native Nanodisc, are routinely used as membrane mimetics to stabilise protein structures in their native conformation. To date, transmembrane proteins of varying complexity (up to 8

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service