Skip to Content
Merck
All Photos(1)

Key Documents

SAE0051

Sigma-Aldrich

Glycerol 3-phosphate Oxidase from Enterococcus sp.

recombinant, expressed in E. coli, lyophilized powder

Synonym(s):

sn-Glycerol 3-phosphate:oxygen 2-oxidoreductase, GPO, L-Glycerol 3-phosphate Oxidase

Sign Into View Organizational & Contract Pricing

Select a Size

500 UNITS
$553.00

$553.00


Check Cart for Availability


Select a Size

Change View
500 UNITS
$553.00

About This Item

EC Number:
UNSPSC Code:
12352204
NACRES:
NA.54

$553.00


Check Cart for Availability

recombinant

expressed in E. coli

Quality Level

form

lyophilized powder

specific activity

≥35 unit/mg solid

storage temp.

−20°C

Related Categories

General description

Glycerol 3-phosphate oxidase is a member of the cytoplasmic oxygen utilizing catabolic enzymes.[1] It is usually a cytoplasmic protein, present in M. pneumoniae. It is produced from glycerophospholipids, that are normally found in the respiratory tract.[2] Glycerol 3-phosphate oxidase helps to reduce dioxygen to water/hydrogen peroxide.[1]

Unit Definition

One unit will oxidize 1.0 micromole of L-glycerol 3-phosphate to dihydroxyacetone phosphate with the formation of H2O2 per min at pH 7.0 and 37 deg C.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Advances in Microbial Physiology: Volume 73, 73 (2018)
Mollicutes: molecular biology and pathogenesis (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service