Skip to Content
Merck
All Photos(2)

Key Documents

SAB5801047

Sigma-Aldrich

Alpaca Anti-Human IgG(Fcγ) Biotin single domain Antibody

recombinant Alpaca(VHH) Nano antibody

Synonym(s):

Recombinant Alpaca single variable domain secondary Nanobody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$795.00

$795.00


Check Cart for Availability


Select a Size

Change View
100 μG
$795.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$795.00


Check Cart for Availability

biological source

alpaca

recombinant

expressed in E. coli

conjugate

biotin

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

recombinant monoclonal

form

lyophilized solid

mol wt

14 kDa

species reactivity

Alpaca, human

technique(s)

ELISA: 1:10000-1:50000
western blot: 1:10000-1:50000

size

0.1 mL

color

colorless

isotype

VHH

shipped in

wet ice

storage temp.

2-8°C

General description

Alpaca Anti-Human IgG(Fcγ) Biotin single domain Antibody is designed to specifically detect human IgG Fcγ fragment. It is a monoclonal, single domain, recombinant antibody to human IgG Fc coupled to Biotin. Based on immunoelectrophoresis and/or ELISA, Alpaca Anti-Human IgG(Fcγ) Biotin single domain Antibody reacts with the Fc fragment of human IgG heavy chain but not with the Fab portion of human IgG.
In mammals, antibodies are classified into five main classes or isotypes – IgA, IgD, IgE, IgG and IgM. They are classed according to the heavy chain they contain – alpha, delta, epsilon, gamma or mu respectively. IgG is the most abundant antibody in normal human serum, accounting for 70-85% of the total immunoglobulin pool. Human IgG consists of four human subclasses (IgG1, IgG2, IgG3 and IgG4), and each contains a different heavy chain. The whole IgG molecule possesses both the Fc region and the Fab region, which possessing the epitope-recognition site. The IgG contains two heavy and light chains(kappa or lambda). The heavy chain is about 50 KD and the light chain is about 25 KD. The common IgG is monomeric with a molecular weight of approximately 150 kD.
VHH are single-domain antibodies derived from the variable regions of heavy chain of Camelidae immunoglobulin. The size of VHH is extremely small(less than15KDa) compared to other forms of antibody fragment, which significantly increase the permeability of VHH. Thus VHH is considered of great value for research, diagnostics and therapeutics.

Specificity

Human IgG(Fcγ fragment specific)
Isotype: VHH domain of alpaca IgG2b/2c

Immunogen

Recombinant Alpaca single variable domain secondary nano antibody

Packaging

100 μg in Glass bottle

Physical form

Lyophilized from 10mM PBS (pH 7.5), 0.05% sucrose, 0.1% trehalose

Reconstitution

Add 100uL dH2O or 100uL dH2O with 50% Glycerol to a final concentration of 1mg/mL

Storage and Stability

After reconstitution, store at –20 °C(Avoid freeze / thaw cycles). Stable for 12 months at -20°C

Other Notes

Recognizes human IgG Fcγ fragment specifically, and reacts with cynomolgus IgG. No Cross-reactivity to rabbit , mouse, rat, goat IgG

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service