Skip to Content
Merck
All Photos(1)

Key Documents

SAB5703014

Sigma-Aldrich

Anti-Butyrylcholinesterase Antibody, clone 4N2N9, Rabbit Monoclonal

Synonym(s):

CHE1, CHE2, E1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$641.00

$641.00


Check Cart for Availability


Select a Size

Change View
100 μL
$641.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

$641.00


Check Cart for Availability

biological source

rabbit

Quality Level

material

colorless

clone

4N2N9, monoclonal

form

liquid

mol wt

90 kDa

species reactivity

human

concentration

1.30 mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

ANFAKYGNPNETQNNSTSWPVFKSTEQKYLTLNTESTRIMTKLRAQQCRFWTSFFPKVLEMTGNIDEAEWEWKAGFHRWNNYMMDWKNQFNDYTSKKESCVGL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BCHE(590)

General description

This gene encodes a cholinesterase enzyme and member of the type-B carboxylesterase/lipase family of proteins. The encoded enzyme exhibits broad substrate specificity and is involved in the detoxification of poisons including organophosphate nerve agents and pesticides, and the metabolism of drugs including cocaine, heroin and aspirin. Humans homozygous for certain mutations in this gene exhibit prolonged apnea after administration of the muscle relaxant succinylcholine. [provided by RefSeq, Jul 2016]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 500-602 of human Butyrylcholinesterase (P06276).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service