Skip to Content
Merck
All Photos(4)

Key Documents

SAB5702906

Sigma-Aldrich

Anti-ACAA2 Antibody, clone 8H6H5, Rabbit Monoclonal

Synonym(s):

ACAA2, DSAEC, acetyl-CoA acyltransferase 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$641.00

$641.00


Check Cart for Availability


Select a Size

Change View
100 μL
$641.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

$641.00


Check Cart for Availability

biological source

rabbit

Quality Level

material

colorless

clone

8H6H5, monoclonal

form

liquid

mol wt

42 kDa

species reactivity

human, rat, rat

concentration

0.5 mg/mL

technique(s)

immunofluorescence: 1:50 - 1:200
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

PAISGALKKAGLSLKDMDLVEVNEAFAPQYLAVERSLDLDISKTNVNGGAIALGHPLGGSGSRITAHLVHELRRRGGKYAVGSACIGGGQGIAVIIQSTA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACAA2(10449)

General description

The encoded protein catalyzes the last step of the mitochondrial fatty acid beta-oxidation spiral. Unlike most mitochondrial matrix proteins, it contains a non-cleavable amino-terminal targeting signal.

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 298-397 of human ACAA2 (P42765).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB, IF/ICC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service