Skip to Content
Merck
All Photos(1)

Key Documents

SAB5702857

Sigma-Aldrich

Anti-HSP105/HSPH1 Antibody, clone 9Y7M10, Rabbit Monoclonal

Synonym(s):

HSP105, HSP105A, HSP105B, NY-CO-25

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$641.00

$641.00


Estimated to ship on01 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$641.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

$641.00


Estimated to ship on01 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

material

colorless

clone

9Y7M10, monoclonal

form

liquid

mol wt

105 kDa

species reactivity

human, rat, rat

concentration

0.5 mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

MSVVGLDVGSQSCYIAVARAGGIETIANEFSDRCTPSVISFGSKNRTIGVAAKNQQITHANNTVSNFKRFHGRAFNDPFIQKEKENLSYDLVPLKNGGVG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HSPH1(10808)

General description

This gene encodes a member of the heat shock protein 70 family of proteins. The encoded protein functions as a nucleotide exchange factor for the molecular chaperone heat shock cognate 71 kDa protein (Hsc70). In addition, this protein plays a distinct but related role as a holdase that inhibits the aggregation of misfolded proteins, including the cystic fibrosis transmembrane conductance regulator (CFTR) protein. Elevated expression of this protein has been observed in numerous human cancers. [provided by RefSeq, Mar 2017]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 1-100 of human HSP105/HSPH1 (Q92598).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service