Skip to Content
Merck
All Photos(1)

Key Documents

SAB5702837

Sigma-Aldrich

Anti-EAAT2/SLC1A2 Antibody, clone 7K5F7, Rabbit Monoclonal

Synonym(s):

EAAT2, EIEE41, GLT-1, HBGT

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$641.00

$641.00


Estimated to ship on15 April 2025



Select a Size

Change View
100 μL
$641.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

$641.00


Estimated to ship on15 April 2025


biological source

rabbit

Quality Level

material

colorless

clone

7K5F7, monoclonal

form

liquid

mol wt

70 kDa

species reactivity

rat, rat, human

concentration

2.50 mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

KKQLGPGKKNDEVSSLDAFLDLIRNLFPENLVQACFQQIQTVTKKVLVAPPPDEEANATSAVVSLLNETVTEVPEETKMVIKKGLEFKDGMNVLGLIGFFI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC1A2(6506)

General description

This gene encodes a member of a family of solute transporter proteins. The membrane-bound protein is the principal transporter that clears the excitatory neurotransmitter glutamate from the extracellular space at synapses in the central nervous system. Glutamate clearance is necessary for proper synaptic activation and to prevent neuronal damage from excessive activation of glutamate receptors. Mutations in and decreased expression of this protein are associated with amyotrophic lateral sclerosis. Alternatively spliced transcript variants of this gene have been identified. [provided by RefSeq, Sep 2010]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 150-250 of human EAAT2/SLC1A2 (P43004).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service