Skip to Content
Merck
All Photos(4)

Key Documents

SAB5702709

Sigma-Aldrich

Anti-FKBP38/FKBP8 Antibody, clone 2J7P6, Rabbit Monoclonal

Synonym(s):

FKBP38, FKBPr38

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$641.00

$641.00


Estimated to ship on30 April 2025



Select a Size

Change View
100 μL
$641.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

$641.00


Estimated to ship on30 April 2025


biological source

rabbit

Quality Level

material

colorless

clone

2J7P6, monoclonal

form

liquid

mol wt

49 kDa

species reactivity

rat, rat, human

concentration

0.40 mg/mL

technique(s)

immunofluorescence: 1:50 - 1:200
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

MASCAEPSEPSAPLPAGVPPLEDFEVLDGVEDAEGEEEEEEEEEEEDDLSELPPLEDMGQPPAEEAEQPGALAREFLAAMEPEPAPAPAPEEWLDILGNG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FKBP8(23770)

General description

The protein encoded by this gene is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. Unlike the other members of the family, this encoded protein does not seem to have PPIase/rotamase activity. It may have a role in neurons associated with memory function. [provided by RefSeq, Jul 2008]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 1-100 of human FKBP38/FKBP8 (Q14318).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB, IF/ICC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service