Skip to Content
Merck
All Photos(1)

Key Documents

SAB5702603

Sigma-Aldrich

Anti-MARK2 Antibody, clone 6O1C2, Rabbit Monoclonal

Synonym(s):

EMK-1, EMK1, PAR-1, Par-1b, Par1b

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$641.00

$641.00


Estimated to ship on30 April 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$641.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

$641.00


Estimated to ship on30 April 2025


Request a Bulk Order

biological source

rabbit

Quality Level

material

colorless

clone

6O1C2, monoclonal

form

liquid

species reactivity

human, rat, rat

concentration

1.90 mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

KPRSLRFTWSMKTTSSMEPNEMMREIRKVLDANSCQSELHEKYMLLCMHGTPGHEDFVQWEMEVCKLPRLSLNGVRFKRISGTSMAFKNIASKIANELKL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MARK2(2011)

General description

This gene encodes a member of the Par-1 family of serine/threonine protein kinases. The protein is an important regulator of cell polarity in epithelial and neuronal cells, and also controls the stability of microtubules through phosphorylation and inactivation of several microtubule-associating proteins. The protein localizes to cell membranes. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Jul 2009]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 689-788 of human MARK2 (Q7KZI7).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service