Skip to Content
Merck
All Photos(1)

Key Documents

SAB5702505

Sigma-Aldrich

Anti-ALAS1 Antibody, clone 2K1G3, Rabbit Monoclonal

Synonym(s):

ALAS, ALAS-H, ALAS3, ALASH, MIG4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$641.00

$641.00


Estimated to ship on10 May 2025



Select a Size

Change View
100 μL
$641.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

$641.00


Estimated to ship on10 May 2025


biological source

rabbit

Quality Level

material

colorless

clone

2K1G3, monoclonal

form

liquid

mol wt

70 kDa

species reactivity

human

concentration

0.58 mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

TEVCDELMSRHNIYVQAINYPTVPRGEELLRIAPTPHHTPQMMNYFLENLLVTWKQVGLELKPHSSAECNFCRRPLHFEVMSEREKSYFSGLSKLVSAQA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ALAS1(211)

General description

This gene encodes the mitochondrial enzyme which is catalyzes the rate-limiting step in heme (iron-protoporphyrin) biosynthesis. The enzyme encoded by this gene is the housekeeping enzyme; a separate gene encodes a form of the enzyme that is specific for erythroid tissue. The level of the mature encoded protein is regulated by heme: high levels of heme down-regulate the mature enzyme in mitochondria while low heme levels up-regulate. A pseudogene of this gene is located on chromosome 12. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq, Jan 2015]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 541-640 of human ALAS1 (P13196).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service