Skip to Content
Merck
All Photos(1)

Key Documents

SAB5702432

Sigma-Aldrich

Anti-Carbonic Anhydrase 1 (CA1) Antibody, clone 0G10F8, Rabbit Monoclonal

Synonym(s):

CA-I, CAB, Car1, HEL-S-11

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$641.00

$641.00


Check Cart for Availability


Select a Size

Change View
100 μL
$641.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$641.00


Check Cart for Availability

biological source

rabbit

Quality Level

material

colorless

clone

0G10F8, monoclonal

form

liquid

mol wt

29 kDa

species reactivity

rat, mouse

concentration

0.75 mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

KVLDALQAIKTKGKRAPFTNFDPSTLLPSSLDFWTYPGSLTHPPLYESVTWIICKESISVSSEQLAQFRSLLSNVEGDNAVPMQHNNRPTQPLKGRTVRASF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CA1(759)

General description

Carbonic anhydrases (CAs) are a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. They participate in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva and gastric acid. They show extensive diversity in tissue distribution and in their subcellular localization. This CA1 gene is closely linked to the CA2 and CA3 genes on chromosome 8. It encodes a cytosolic protein that is found at the highest level in erythrocytes. Allelic variants of this gene have been described in some populations. Alternative splicing and the use of alternative promoters results in multiple transcript variants. [provided by RefSeq, Nov 2016]

Immunogen

PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.
Recombinant fusion protein containing a sequence corresponding to amino acids 160-261 of human Carbonic Anhydrase 1 (CA1) (CA1) (P00915).

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service