Skip to Content
Merck
All Photos(4)

Key Documents

SAB5702396

Sigma-Aldrich

Anti-DCR2 Antibody, clone 10Y1N4, Rabbit Monoclonal

Synonym(s):

CD264, DCR2, TRAIL-R4, TRAILR4, TRUNDD

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$641.00

$641.00


Estimated to ship on01 May 2025



Select a Size

Change View
100 μL
$641.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

$641.00


Estimated to ship on01 May 2025


biological source

rabbit

Quality Level

material

colorless

clone

10Y1N4, monoclonal

form

liquid

mol wt

45-60 kDa

species reactivity

rat, rat, human

concentration

0.61 mg/mL

technique(s)

immunohistochemistry: 1:50 - 1:200
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

PTQVSEQEIQGQELAELTGVTVELPEEPQRLLEQAEAEGCQRRRLLVPVNDADSADISTLLDASATLEEGHAKETIQDQLVGSEKLFYEEDEAGSATSCL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor contains an extracellular TRAIL-binding domain, a transmembrane domain, and a truncated cytoplamic death domain. This receptor does not induce apoptosis, and has been shown to play an inhibitory role in TRAIL-induced cell apoptosis. [provided by RefSeq, Jul 2008]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 287-386 of human DCR2 (Q9UBN6).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB, IHC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service