Skip to Content
Merck
All Photos(6)

Key Documents

SAB5702377

Sigma-Aldrich

Anti-Laminin beta 1 (LAMB1) Antibody

rabbit monoclonal, 6I9T4

Synonym(s):

CLM, LIS5

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$641.00

$641.00


Estimated to ship on16 April 2025



Select a Size

Change View
100 μL
$641.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

$641.00


Estimated to ship on16 April 2025


Product Name

Anti-Laminin beta 1 Antibody, clone 6I9T4, Rabbit Monoclonal,

biological source

rabbit

Quality Level

material

colorless

clone

6I9T4, monoclonal

form

liquid

mol wt

250 kDa

species reactivity

rat, human, rat

concentration

1 mg/mL

technique(s)

immunofluorescence: 1:50 - 1:200
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

KKTLDGELDEKYKKVENLIAKKTEESADARRKAEMLQNEAKTLLAQANSKLQLLKDLERKYEDNQRYLEDKAQELARLEGEVRSLLKDISQKVAVYSTCL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LAMB1(3912)

General description

Laminins, a family of extracellular matrix glycoproteins, are the major noncollagenous constituent of basement membranes. They have been implicated in a wide variety of biological processes including cell adhesion, differentiation, migration, signaling, neurite outgrowth and metastasis. Laminins are composed of 3 non identical chains: laminin alpha, beta and gamma and they form a cruciform structure consisting of 3 short arms, each formed by a different chain, and a long arm composed of all 3 chains. Different alpha, beta and gamma chain isomers combine to give rise to different heterotrimeric laminin isoforms which are designated by Arabic numerals in the order of their discovery, i.e. alpha1beta1gamma1 heterotrimer is laminin 1. This gene encodes the beta chain isoform laminin, beta 1. The beta 1 chain has 7 structurally distinct domains which it shares with other beta chain isomers. The C-terminal helical region containing domains I and II are separated by domain alpha, domains III and V contain several EGF-like repeats, and domains IV and VI have a globular conformation. Laminin, beta 1 is expressed in most tissues that produce basement membranes, and is one of the 3 chains constituting laminin 1, the first laminin isolated from Engelbreth-Holm-Swarm (EHS) tumor. A sequence in the beta 1 chain that is involved in cell attachment, chemotaxis, and binding to the laminin receptor was identified and shown to have the capacity to inhibit metastasis.

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 1687-1786 of human Laminin beta 1 (P07942).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB, IF/ICC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service