Skip to Content
Merck
All Photos(1)

Key Documents

SAB5702267

Sigma-Aldrich

Anti-PQBP1 Antibody, clone 8D8H6, Rabbit Monoclonal

Synonym(s):

PQBP1,MRX2,MRX55,MRXS3,MRXS8,NPW38,RENS1,SHS

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$641.00

$641.00


Check Cart for Availability


Select a Size

Change View
100 μL
$641.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

$641.00


Check Cart for Availability

biological source

rabbit

Quality Level

material

colorless

clone

8D8H6, monoclonal

form

liquid

mol wt

38 kDa

species reactivity

rat, mouse

concentration

2.00 mg/mL

technique(s)

western blot: 1:200 - 1:2000

color

colorless

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PQBP1(10084)

General description

This gene encodes a nuclear polyglutamine-binding protein that is involved with transcription activation. The encoded protein contains a WW domain. Mutations in this gene have been found in patients with Renpenning syndrome 1 and other syndromes with X-linked mental retardation. Multiple alternatively spliced transcript variants that encode different protein isoforms have been described for this gene.

Immunogen

PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.
Recombinant protein of human PQBP1.

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service