Skip to Content
Merck
All Photos(1)

Documents

SAB5702266

Sigma-Aldrich

Anti-SLFN11 Antibody, clone 8G3G0, Rabbit Monoclonal

Synonym(s):

SLFN8/9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

8G3G0, monoclonal

form

liquid

mol wt

101 kDa

species reactivity

human

concentration

1 mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

LYRRSETSVRSMDSREAFCFLKTKRKPKILEEGPFHKIHKGVYQELPNSDPADPNSDPADLIFQKDYLEYGEILPFPESQLVEFKQFSTKHFQEYVKRTIPEYVPAFANTGGGYLFIGVDDKSREVLGCAKENVDPDSLRRKIEQAIYKLPCVHFCQPQRPITFTLKIVNVLKRGELYGYACMIRVNPFCCAVFSEA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLFN11(91607)

General description

Inhibitor of DNA replication that promotes cell death in response to DNA damage. Acts as a guardian of the genome by killing cells with defective replication. Persistently blocks stressed replication forks by opening chromatin across replication initiation sites at stressed replication forks, possibly leading to unwind DNA ahead of the MCM helicase and block fork progression, ultimately leading to cell death. Acts independently of ATR. Also acts as an interferon (IFN-induced antiviral protein which acts as an inhibitor of retrovirus protein synthesis. Specifically abrogates the production of retroviruses such as human immunodeficiency virus 1 (HIV-1 by acting as a specific inhibitor of the synthesis of retroviruses encoded proteins in a codon-usage-dependent manner. Binds to tRNAs and exploits the unique viral codon bias towards A/T nucleotides. The exact inhibition mechanism is unclear: may either sequester tRNAs, prevent their maturation via post-transcriptional processing or may accelerate their deacylation. Does not inhibit reverse transcription, integration or production and nuclear export of viral RNA.

Immunogen

PBS with 0.05% proclin300,0.05% BSA,50% glycerol,pH7.3.
Recombinant fusion protein containing a sequence corresponding to amino acids 132-328 of human SLFN11 (NP_689483.3).

Application

WB

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service