Skip to Content
Merck
All Photos(3)

Documents

SAB5702236

Sigma-Aldrich

Anti-SATB2 Antibody, clone 3F9E1, Rabbit Monoclonal

Synonym(s):

SATB2,GLSS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

3F9E1, monoclonal

form

liquid

mol wt

100 kDa

species reactivity

mouse, mouse

concentration

0.55 mg/mL

technique(s)

immunofluorescence: 1:50 - 1:100
immunohistochemistry: 1:1000 - 1:2500
immunoprecipitation (IP): 1:50 - 1:200
western blot: 1:10000 - 1:11000

color

colorless

isotype

IgG

immunogen sequence

HDVGLYPDQEAIHTLSAQLDLPKHTIIKFFQNQRYHVKHHGKLKEHLGSAVDVAEYKDEELLTESEENDSEEGSEEMYKVEAEEENADKSKAAPAEIDQR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SATB2(23314)

General description

This gene encodes a DNA binding protein that specifically binds nuclear matrix attachment regions. The encoded protein is involved in transcription regulation and chromatin remodeling. Defects in this gene are associated with isolated cleft palate and cognitive disability. Alternate splicing results in multiple transcript variants that encode the same protein.

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 634-733 of human SATB2 (NP_001165980.1).
PBS with 0.05% proclin300,0.05% BSA,50% glycerol,pH7.3.

Application

WB, IHC, IF/ICC, IP

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service