Skip to Content
Merck
All Photos(1)

Key Documents

SAB5702161

Sigma-Aldrich

Anti-CHX10 Antibody, clone 4Z4R2, Rabbit Monoclonal

Synonym(s):

CHX10, HOX10, MCOP2, MCOPCB3, RET1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$641.00

$641.00


Check Cart for Availability


Select a Size

Change View
100 μL
$641.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

$641.00


Check Cart for Availability

biological source

rabbit

Quality Level

material

colorless

clone

4Z4R2, monoclonal

form

liquid

mol wt

39 kDa

species reactivity

rat, human, rat

concentration

3.00 mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

AAAESGRKPEGERQALPKLDKMEQDERGPDAQAAISQEELRENSIAVLRAKAQEHSTKVLGTVSGPDSLARSTEKPEEEEAMDEDRPAERLSPPQLEDMA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VSX2(338917)

General description

This gene encodes a homeobox protein originally described as a retina-specific transcription factor. Mutations in this gene are associated with microphthalmia, cataracts and iris abnormalities. [provided by RefSeq, Oct 2009]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 262-361 of human CHX10 (P58304).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service