Skip to Content
Merck
All Photos(1)

Key Documents

SAB5702129

Sigma-Aldrich

Anti-LGI1 Antibody, clone 2P8Z9, Rabbit Monoclonal

Synonym(s):

LGI1, ADLTE, ADPAEF, ADPEAF, EPITEMPIN, EPT, ETL1, IB1099, leucine rich glioma inactivated 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$641.00

$641.00


Estimated to ship on21 May 2025



Select a Size

Change View
100 μL
$641.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

$641.00


Estimated to ship on21 May 2025


biological source

rabbit

Quality Level

material

colorless

clone

2P8Z9, monoclonal

form

liquid

mol wt

64 kDa

species reactivity

rat, human, rat

concentration

0.67 mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

NLQTLPKDIFKGLDSLTNVDLRGNSFNCDCKLKWLVEWLGHTNATVEDIYCEGPPEYKKRKINSLSSKDFDCIITEFAKSQDLPYQSLSIDTFSYLNDEYV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... Lgi1(9211)

General description

This gene encodes a member of the secreted leucine-rich repeat (LRR) superfamily and shares homology with members of the SLIT protein family. The encoded protein may regulate the activity of voltage-gated potassium channels and may be involved in neuronal growth regulation and cell survival. This gene is rearranged as a result of translocations in glioblastoma cell lines, and it is frequently down-regulated or rearranged in malignant gliomas. Mutations in this gene result in autosomal dominant lateral temporal epilepsy. Alternative splicing results in multiple transcript variants.

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 150-250 of human LGI1 (O95970).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service