Skip to Content
Merck
All Photos(1)

Key Documents

SAB5702123

Sigma-Aldrich

Anti-ATG12 Antibody, clone 9R8W2, Rabbit Monoclonal

Synonym(s):

ATG12,APG12,APG12L,FBR93,HAPG12,Apg12

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$641.00

$641.00


Check Cart for Availability


Select a Size

Change View
100 μL
$641.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

$641.00


Check Cart for Availability

biological source

rabbit

Quality Level

material

colorless

clone

9R8W2, monoclonal

form

liquid

mol wt

55 kDa

species reactivity

rat, rat, human

concentration

2 mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

AVGDTPIMKTKKWAVERTRTIQGLIDFIKKFLKLVASEQLFIYVNQSFAPSPDQEVGTLYECFGSDGKLVLHYCKSQAWG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ATG12(9140)

General description

Autophagy is a process of bulk protein degradation in which cytoplasmic components, including organelles, are enclosed in double-membrane structures called autophagosomes and delivered to lysosomes or vacuoles for degradation. ATG12 is the human homolog of a yeast protein involved in autophagy (Mizushima et al., 1998 [PubMed 9852036]).[supplied by OMIM, Mar 2008]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 61-140 of human Apg12 (O94817).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service