Skip to Content
Merck
All Photos(2)

Key Documents

SAB5701966

Sigma-Aldrich

Anti-RANBP3 Antibody, clone 8H7W3, Rabbit Monoclonal

Synonym(s):

RANBP3, ran-binding protein 3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$641.00

$641.00


Estimated to ship on27 April 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$641.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

$641.00


Estimated to ship on27 April 2025


Request a Bulk Order

biological source

rabbit

Quality Level

material

colorless

clone

8H7W3, monoclonal

form

liquid

species reactivity

rat, mouse

concentration

3.00 mg/mL

technique(s)

immunohistochemistry: 1:50 - 1:200

color

colorless

isotype

IgG

immunogen sequence

MADLANEEKPAIAPPVFVFQKDKGQKSPAEQKNLSDSGEEPRGEAEAPHHGTGHPESAGEHALEPPAPAGASASTPPPPAPEAQLPPFPRELAGRSAGGS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RANBP3(8498)

General description

This gene encodes a protein with a RanBD1 domain that is found in both the nucleus and cytoplasm. This protein plays a role in nuclear export as part of a heteromeric complex. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 1-100 of human RANBP3 (Q9H6Z4).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

IHC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service