Skip to Content
Merck
All Photos(1)

Key Documents

SAB5700411

Sigma-Aldrich

Anti-Phospho-PDGFRA-Y849 antibody produced in rabbit

Synonym(s):

CD140A, GAS9, PDGFR-2, PDGFR2, PDGFRA, RHEPDGFRA

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$724.00

$724.00


Check Cart for Availability


Select a Size

Change View
100 μL
$724.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$724.00


Check Cart for Availability

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

122
24
82

species reactivity

human

concentration

2.21 mg/ml

technique(s)

western blot: 1:500-1:2000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr849)

Gene Information

human ... PDGFRA(5156)

General description

This gene encodes a cell surface tyrosine kinase receptor for members of the platelet-derived growth factor family. These growth factors are mitogens for cells of mesenchymal origin. The identity of the growth factor bound to a receptor monomer determines whether the functional receptor is a homodimer or a heterodimer, composed of both platelet-derived growth factor receptor alpha and beta polypeptides. Studies suggest that this gene plays a role in organ development, wound healing, and tumor progression. Mutations in this gene have been associated with idiopathic hypereosinophilic syndrome, somatic and familial gastrointestinal stromal tumors, and a variety of other cancers.

Immunogen

A synthetic phosphorylated peptide around Y849 of human PDGFRA (NP_006197.1).

Physical form

PBS with 0.02% sodium azide,50% glycerol,pH7.3.

Storage and Stability

Store at -20℃. Avoid freeze / thaw cycles.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service