Skip to Content
Merck
All Photos(6)

Key Documents

SAB5700096

Sigma-Aldrich

Anti-CYP3A4 antibody produced in rabbit

Synonym(s):

CP33, CP34, CYP3A, CYP3A3, CYP3A4, CYPIIIA3, CYPIIIA4, HLP, NF-25, P450C3, P450PCN1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$672.00

$672.00


Check Cart for Availability


Select a Size

Change View
100 μL
$672.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$672.00


Check Cart for Availability

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

57

species reactivity

human, rat, mouse

concentration

0.77 mg/ml

technique(s)

immunofluorescence: 1:20-1:100
immunohistochemistry: 1:20-1:50
western blot: 1:500-1:1000

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CYP3A4(1576)

General description

This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases that catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and its expression is induced by glucocorticoids and some pharmacological agents. This enzyme is involved in the metabolism of approximately half the drugs in use today, including acetaminophen, codeine, cyclosporin A, diazepam and erythromycin. The enzyme also metabolizes some steroids and carcinogens. This gene is part of a cluster of cytochrome P450 genes on chromosome 7q21.1. Previously another CYP3A gene, CYP3A3, was thought to exist; however, it is now thought that this sequence represents a transcript variant of CYP3A4. Alternatively spliced transcript variants encoding different isoforms have been identified.

Immunogen

Recombinant fusion protein containing a sequence corresponding to amino acids 244-503 of human CYP3A4 (NP_059488.2).

Physical form

PBS with 0.02% sodium azide,pH7.3.

Storage and Stability

Store at 4℃. Avoid freeze / thaw cycles.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service