Skip to Content
Merck
All Photos(9)

Key Documents

SAB5700016

Sigma-Aldrich

Anti-PARP1 antibody produced in rabbit

Synonym(s):

ADPRT, ADPRT 1, ADPRT1, ARTD1, Cleaved PARP, PARP, PARP-1, PARP1, PPOL, pADPRT-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$615.00

$615.00


Check Cart for Availability


Select a Size

Change View
100 μL
$615.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$615.00


Check Cart for Availability

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

113

species reactivity

rat, human, mouse

concentration

2.78 mg/ml

technique(s)

ChIP: 1:50-1:200
immunofluorescence: 1:50-1:200
immunohistochemistry: 1:50-1:200
immunoprecipitation (IP): 1:50-1:200
western blot: 1:1000-1:3000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PARP1(142)

General description

This gene encodes a chromatin-associated enzyme, poly(ADP-ribosyl)transferase, which modifies various nuclear proteins by poly(ADP-ribosyl)ation. The modification is dependent on DNA and is involved in the regulation of various important cellular processes such as differentiation, proliferation, and tumor transformation and also in the regulation of the molecular events involved in the recovery of cell from DNA damage. In addition, this enzyme may be the site of mutation in Fanconi anemia, and may participate in the pathophysiology of type I diabetes.

Immunogen

Recombinant fusion protein containing a sequence corresponding to amino acids 81-390 of human PARP1 (NP_001609.2).

Physical form

PBS with 0.02% sodium azide,50% glycerol,pH7.3.

Storage and Stability

Store at -20℃. Avoid freeze / thaw cycles.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service