Skip to Content
Merck
All Photos(2)

Key Documents

SAB5600252

Sigma-Aldrich

Anti-Cytokeratin 19 (CK-19) antibody, Rabbit monoclonal

recombinant, expressed in HEK 293 cells, clone RM364, purified immunoglobulin

Synonym(s):

Keratin 19

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$730.00
1 ML
Inquire

$730.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1599


Select a Size

Change View
100 μL
$730.00
1 ML
Inquire

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$730.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1599

recombinant

expressed in HEK 293 cells

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RM364, monoclonal
recombinant monoclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:500—1:1000
western blot: 1:1000-1:2000

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KRT19(3880)

Specificity

This antibody reacts human CK-19 (Cytokeratin-19).

Immunogen

A peptide corresponding to the C-terminus of human CK-19 (Cytokeratin-19)

Physical form

Solution in phosphate buffered saline containing 50% Glycerol/PBS with 1% BSA and 0.09% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service