Skip to Content
Merck
All Photos(3)

Key Documents

SAB5600004

Sigma-Aldrich

Anti-Mouse IgG2c antibody, Rabbit monoclonal

recombinant, expressed in HEK 293 cells, clone RM223, purified immunoglobulin

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$779.00
1 MG
Inquire

$779.00


Check Cart for Availability


Select a Size

Change View
100 μG
$779.00
1 MG
Inquire

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

$779.00


Check Cart for Availability

recombinant

expressed in HEK 293 cells

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RM223, monoclonal
recombinant monoclonal

form

buffered aqueous glycerol solution

species reactivity

mouse

concentration

~1 mg/mL

technique(s)

ELISA: 0.01 μg/mL
immunoblotting: 0.5-2 μg/mL (non-reduced only)
immunocytochemistry: 0.5-2 μg/mL
immunohistochemistry: 0.5-2 μg/mL

isotype

IgG

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

IgG2c codes for Igh-1b. In mice, IgG2a and IgG2c are arranged in tandem on identical chromosome as two visible loci.

Specificity

This antibody reacts to the Fc region of mouse IgG2c. No cross reactivity with mouse IgG1, IgG2a, IgG2b, IgG3, IgM, IgA, IgE, human IgG, or rat IgG.

Immunogen

Purified mouse IgG2c whole molecule

Biochem/physiol Actions

Mouse IgG2c generate several unique non-bonded interactions between the BC, DE and FG loops than mIgG2b. It connects to FcγRIV (Fc γ receptor IV) with a high affinity in the complex-type glycoform. In mice, IgG2c has unique immune profiles like, response, immune-decoration, because of their difference in protein sequence.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline containing 50% glycerol, 1% BSA and 0.09% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Possible allelic structure of IgG2a and IgG2c in mice.
Zhang Z, et al.
Molecular Immunology, 50(3), 169-171 (2012)
Mouse IgG2c Fc loop residues promote greater receptor-binding affinity than mouse IgG2b or human IgG1.
Daniel JF, et al.
PLoS ONE, 13(2), e0192123-e0192123 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service