Skip to Content
Merck
All Photos(2)

Key Documents

SAB5500090

Sigma-Aldrich

Anti-Cyclin D1 antibody, Rabbit monoclonal

clone SP4, recombinant, expressed in proprietary host, tissue culture supernatant

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$469.00

$469.00


Check Cart for Availability


Select a Size

Change View
100 μL
$469.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$469.00


Check Cart for Availability

biological source

rabbit

Quality Level

recombinant

expressed in proprietary host

conjugate

unconjugated

antibody form

tissue culture supernatant

antibody product type

primary antibodies

clone

SP4, monoclonal

species reactivity

human (tested)

species reactivity (predicted by homology)

frog, dog, mouse, chicken, rat, bovine

technique(s)

immunoblotting: 1:25
immunohistochemistry: 1:100

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CCND1(595)

General description

Cyclin D1 or PRAD-1 or bcl-1 is one of the key cell cycle regulators, and functions in association with cdk4 and / or cdk6 by phosphorylating the Rb protein. It is a putative proto-oncogene overexpressed in a wide variety of human neoplasms including mantle cell lymphomas (MCL).

Immunogen

Synthetic peptide from C-terminus of human cyclin D1.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

0.1 ml rabbit monoclonal antibody supplied as tissue culture supernatant in TBS/1% BSA buffer pH 7.5 with less than 0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isabel Leung et al.
Blood advances, 7(12), 2718-2730 (2022-12-06)
Therapy with CD19-directed chimeric antigen receptor (CAR) T cells has transformed the treatment of advanced B-cell malignancies. However, loss of or low antigen expression can enable tumor escape and limit the duration of responses achieved with CAR T-cell therapy. Engineering
Cindy G Avalos-de León et al.
Cells, 8(12) (2019-12-15)
In liver transplantation (LT), organ shortage has led to the use of steatotic and non-steatotic grafts from donors after cardiocirculatory death (DCD). However, these grafts, especially those with steatosis, exhibit poor post-operative outcomes. To address this problem, we investigated the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service