SAB5202150
Monoclonal Anti-GABA A Receptor-Atto 488 antibody produced in mouse
clone S398A-34
Select a Size
$354.00
Select a Size
About This Item
$354.00
biological source
mouse
conjugate
Atto 488 conjugate
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
S398A-34, monoclonal
form
buffered aqueous solution
mol wt
60 kDa
species reactivity
human, mouse
concentration
1.0 mg/mL
technique(s)
immunofluorescence: 1:100
western blot: 1:1000
isotype
IgG1
NCBI accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
rat ... Gabra4(140675)
1 of 4
This Item | SAB5202239 | SAB5202095 | SAB5202160 |
---|---|---|---|
species reactivity human, mouse | species reactivity human, mouse | species reactivity human, rat | species reactivity human, mouse |
antibody form purified immunoglobulin | antibody form purified immunoglobulin | antibody form purified immunoglobulin | antibody form purified immunoglobulin |
Gene Information rat ... Gabra4(140675) | Gene Information human ... GABRA5(2558) | Gene Information rat ... Gabra2(289606) | Gene Information rat ... Gabra4(140675) |
conjugate Atto 488 conjugate | conjugate Atto 390 conjugate | conjugate Atto 390 conjugate | conjugate FITC conjugate |
clone S398A-34, monoclonal | clone S415-24, monoclonal | clone S399-19, monoclonal | clone S398A-34, monoclonal |
Immunogen
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service