Skip to Content
Merck
All Photos(5)

Documents

SAB5201881

Sigma-Aldrich

Monoclonal Anti-HSP70-Atto 595 antibody produced in mouse

clone 1H11

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.45

biological source

mouse

conjugate

(Atto 595 Conjugate)

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1H11, monoclonal

form

buffered aqueous solution

mol wt

70 kDa

species reactivity

human

concentration

1.0 mg/mL

technique(s)

flow cytometry: 1:250
immunofluorescence: 1:100
western blot: 1:1000

isotype

IgG1

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... HSPA1A(3303)

Immunogen

Human native HSP70 protein

Physical form

PBS pH 7.4, 50% glyercol, 0.09% Sodium Azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service