Skip to Content
Merck
All Photos(1)

Key Documents

SAB5200204

Sigma-Aldrich

Monoclonal Anti-Hsp70 antibody produced in rat

clone 7FB, purified immunoglobulin

Synonym(s):

Anti-Dm-hsp70, Anti-Hsp70Bb, Anti-dHsp70

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$666.00

$666.00


Check Cart for Availability


Select a Size

Change View
100 μG
$666.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$666.00


Check Cart for Availability

biological source

rat

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7FB, monoclonal

form

buffered aqueous solution

mol wt

antigen predicted mol wt 70 kDa

species reactivity

Drosophila

concentration

1 mg/mL

technique(s)

ELISA: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG2b

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Drosophila melanogaster ... Hsp70Bb(48582)

Specificity

Detects heat-inducible form of HSP70, ~70kDa.

Immunogen

Prepared from Drosophila tissue culture cells heat shocked at 36.5◦C for 3 hours, and isolated using SDS PAGE.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

PBS pH7.4, 50% glycerol, 0.1% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Max Jameson Aragon et al.
eLife, 11 (2022-01-25)
We developed a multiphoton imaging method to capture neural structure and activity in behaving flies through the intact cuticle. Our measurements showed that the fly head cuticle has surprisingly high transmission at wavelengths >900nm, and the difficulty of through-cuticle imaging

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service