Skip to Content
Merck
All Photos(3)

Key Documents

SAB5200084

Sigma-Aldrich

Anti-CANX antibody produced in rabbit

whole antiserum

Synonym(s):

Anti-CANX, Anti-CNX, Anti-Calnexin -NT, Anti-IP90, Anti-P90

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$482.00

$482.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1147


Select a Size

Change View
200 μL
$482.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$482.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1147

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

form

aqueous solution

mol wt

antigen predicted mol wt 90 kDa

species reactivity

rabbit, sheep, Xenopus, mouse, rat, bovine, hamster, dog, human, guinea pig, monkey, chicken, pig

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

canine ... CANX(403908)

General description

Calnexin (CANX) is a 67kDa transmembrane protein expressed in the endoplasmic reticulum. It is part of the lectin chaperone group. The gene ID of the protein is 821.

Specificity

Detects ~90 kDa.

Immunogen

A 19 residue synthetic peptide based on canine calnexin and the peptide coupled to KLH.

Biochem/physiol Actions

Calnexin (CANX) takes care of protein folding. It binds to the target proteins which have only one glucose residue and aids in their folding. When the protein is released from CANX, it is then trafficked to the Golgi apparatus for further modifications.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit Antiserum

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetyana Drozdova et al.
Physiological reports, 1(4), e00086-e00086 (2013-12-05)
Nephrin, an important component of the podocyte filtration slit diaphragm, plays a key role in the maintenance of glomerular permselectivity. Mutations in nephrin lead to proteinuria and congenital nephrotic syndrome. Nephrin undergoes posttranslational modifications in the endoplasmic reticulum (ER) prior
Dorthe T Olsen et al.
Protein expression and purification, 92(1), 105-111 (2013-09-24)
Calreticulin (Crt) and calnexin (Cnx) are homologous endoplasmic reticulum (ER) chaperones involved in protein folding and quality control. Crt is a soluble ER luminal Mr 46 kDa protein and Cnx is a Mr 67kDa ER membrane protein. During purification of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service