SAB5200003
Monoclonal Anti-DLG4 antibody produced in mouse
clone 7E3, 1 mg/mL, purified immunoglobulin
Synonym(s):
Anti-DLG4, Anti-PSD95, Anti-SAP90
About This Item
Recommended Products
biological source
mouse
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
7E3, monoclonal
form
buffered aqueous glycerol solution
mol wt
antigen predicted mol wt 100 kDa
species reactivity
mouse, rat, bovine
concentration
1 mg/mL
technique(s)
western blot: suitable
isotype
IgG2a
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
rat ... Dlg4(29495)
Related Categories
Specificity
Immunogen
Features and Benefits
Physical form
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service