Skip to Content
Merck
All Photos(1)

Key Documents

SAB4700972

Sigma-Aldrich

Monoclonal Anti-MAP2 antibody produced in mouse

clone MT-01

Synonym(s):

MAP-2, Microtubule-associated protein 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$576.00

$576.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZMS1013


Select a Size

Change View
100 μG
$576.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$576.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZMS1013

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

MT-01, monoclonal

form

buffered aqueous solution

species reactivity

mouse, pig, human, bovine, cat

concentration

1 mg/mL

technique(s)

western blot: 1 μg/mL

accession no.

P11137

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... MAP2(4133)

General description

The antibody MT-01 reacts with molecule Microtubule Associated Protein 2ab (MAP2ab).

Immunogen

Termostabile fraction of bovine brain microtubules (whole MAP2ab molecule).

Physical form

Solution in phosphate buffered saline with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service