Skip to Content
Merck
All Photos(2)

Key Documents

SAB4700749

Sigma-Aldrich

Monoclonal Anti-PRR7 / TRAP3 antibody produced in mouse

clone TRAP3/10, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-TRAP3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$257.00

$257.00


Check Cart for Availability


Select a Size

Change View
100 μG
$257.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$257.00


Check Cart for Availability

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

TRAP3/10, monoclonal

form

buffered aqueous solution

species reactivity

human, mouse, rat

concentration

1 mg/mL

technique(s)

immunoblotting: suitable
immunocytochemistry: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... PRR7(80758)

General description

The mouse monoclonal antibody TRAP3/10 recognizes an epitope located in the C-terminal part of the intracellular domain of PRR7/TRAP3 (amino acids 126-253 of human PRR7 / TRAP3), a 28 kDa proline-rich membrane protein presumably associated with NMDA receptor complex.

Immunogen

Recombinant C-terminal half of the intracellular domain of human PRR7/TRAP3 (amino acids 126-253)

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service