Skip to Content
Merck
All Photos(1)

Key Documents

SAB4700735

Sigma-Aldrich

Monoclonal Anti-CD205 antibody produced in mouse

clone HD30, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-DEC-205

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$292.00

$292.00


Check Cart for Availability


Select a Size

Change View
100 μG
$292.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$292.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

HD30, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

1 mg/mL

technique(s)

flow cytometry: suitable
immunoblotting: suitable
immunohistochemistry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... LY75(4065)

General description

The mouse monoclonal antibody HD30 recognizes CD205, an approx. 200 kDa C-type lectin transmembrane protein of the MMR (macrophage mannose receptor) family, expressed at high levels on dendritic cells and thymic epithelial cells, and at low levels on lymphocytes, NK cells and monocytes.

Immunogen

Recombinant Fc-tagged human CD205

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service