Skip to Content
Merck
All Photos(1)

Key Documents

SAB4700517

Sigma-Aldrich

Monoclonal Anti-CD71-Biotin antibody produced in mouse

clone MEM-75, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-TFRC, Anti-Transferrin Receptor

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$335.00

$335.00


Estimated to ship on07 May 2025



Select a Size

Change View
100 μG
$335.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$335.00


Estimated to ship on07 May 2025


biological source

mouse

Quality Level

conjugate

biotin conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

MEM-75, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

1 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... TFRC(7037)

Related Categories

General description

The antibody MEM-75 reacts with CD71 antigen (transferrin receptor), a 95 kDa type II homodimeric transmembrane glycoprotein expressed on activated B and T lymphocytes, macrophages and erythroid precursors; it is lost on resting blood leukocytes.

Immunogen

NALM-6 human pre-B cell line

Application

The reagent is designed for Flow Cytometry analysis. Suggested working dilution is 1:200. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroko Sugiura et al.
The Journal of biological chemistry, 279(22), 23343-23348 (2004-04-07)
Endophilin 1 (Endo1) participates in synaptic vesicle biogenesis through interactions of its Src homology 3 domain with the polyphosphoinositide phosphatase Synaptojanin and the GTPase Dynamin. Endo1 has also been reported to affect endocytosis by converting membrane curvature via its lysophosphatidic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service